NSI Lab Solutions 9001-10 E. coli High Range NCTC 9001 Environmental Culture; 10/PK
Representative image only.
NSI Lab Solutions 9001-10 E. coli High Range NCTC 9001 Environmental Culture; 10/PK
NSI Lab Solutions | Item: EW-05002-28
A single pellet contains a pure culture that will yield a specific certified activity level—activity level is stated on the product lot COA
$175.00USD / Pkg of 10
$175.00USD / Pkg of 10
Product Description
Key Features
  • Hydration fluid dissolves culture pellet in less than a minute
  • No dilution needed—hydrate then directly inoculate media
  • Less than 10% RSD lot precision
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • No royalty fee
  • Pack of 10
More About this Item
These microbiological reference materials are designed for use as quantitative control for environmental water analysis. To use, simply transfer the pellet to a sterile room temperature hydration fluid. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Directly inoculate media without serial dilution for convenience.
Use E. coli NCTC 9001 culture as a quantitative control for water coliform analyses. The Certificate of Analysis (CoA) lists value assignments for six water coliform test parameters: Total coliform, Fecal Coliform, and E. coli by both MPN methods and SM plate count methods. Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
NCTC reference cultures obtained from the National Collection of Type Cultures.
Specifications
  • OrganismE. coli
  • Packaging10/PK
  • Product TypeEnvironmental Single Cultures
  • Shelf Life Months (months)18
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • StrainNCTC 9001, ATCC 1175
  • DescriptionE. coli High Range NCTC 9001 Environmental Culture; 10/Pk