NSI Lab Solutions 10006-10 E. aerogenes High Range NCTC 10006 Environmental Culture; 10/PK
Representative image only.
NSI Lab Solutions 10006-10 E. aerogenes High Range NCTC 10006 Environmental Culture; 10/PK
NSI Lab Solutions | Item: EW-61203-58
A single pellet contains a pure culture that will yield a specific certified activity level—activity level is stated on the product lot COA
$175.00USD / Pkg of 10
$175.00USD / Pkg of 10
Product Description
Key Features
  • Hydration fluid dissolves culture pellet in less than a minute
  • No dilution needed—hydrate then directly inoculate media
  • Less than 10% RSD lot precision
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • No royalty fee
  • Pack of 10
More About this Item
These microbiological reference materials are designed for use as quantitative control for environmental water analysis. To use, simply transfer the pellet to a sterile room temperature hydration fluid. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Directly inoculate media without serial dilution for convenience.
Use E. aerogenes NCTC 10006 culture as a total coliform positive control for environmental water analysis. The Certificate of Analysis (CoA) lists value assignments for MPN methods and plate count methods. Store in the freezer.
Specifications
  • OrganismE. aerogenes
  • Packaging10/PK
  • Product TypeEnvironmental Single Cultures
  • Shelf Life Months (months)18
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • StrainNCTC 10006, ATCC 13048
  • DescriptionE. aerogenes High Range NCTC 10006 Environmental Culture; 10/Pk