NSI Lab Solutions CMPT-081B Quantitative Water Activity in Oil PT Express™; 2/PK
Representative image only.
NSI Lab Solutions CMPT-081B Quantitative Water Activity in Oil PT Express™; 2/PK
NSI Lab Solutions | Item: EW-61204-66
On-demand proficiency test (PT) for when you need results fast
$373.00USD / Pkg of 2
$373.00USD / Pkg of 2
Product Description
Key Features
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • For quantitative determination of water activity in hemp oil
  • Supplied in duplicate for convenience and value
More About this Item
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results. Your PT report is available within 48 hours once you submit your results online.
These PTs are suitable for compliance with BCC and Florida as well as other USA states' statutory requirements.
Hemp PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
Specifications
  • Packaging2/PK
  • Product TypeHemp Chemistry Proficiency Test
  • Shelf Life Months (months)36
  • Shipping requirementFrozen ice packs
  • Storage requirementRefrigerate between 2 and 8°C
  • DescriptionQuantitative Water Activity in Oil PT Express™; 2/PK