NSI Lab Solutions FMPT-024D Qualitative STEC with Dairy Matrix Proficiency Test Set; 3/PK

Representative image only.

NSI Lab Solutions FMPT-024D Qualitative STEC with Dairy Matrix Proficiency Test Set; 3/PK

NSI Lab Solutions | Item: EW-05004-02

Ensure state and accreditation regulation compliance with proficiency testing

$265.00USD / Pkg of 3

$265.00USD / Pkg of 3
Product Description

Includes

Three jars of sterile skim milk powder matrix and three individually packaged lyophilized microorganism pellets.

Key Features

  • Proficiency tests for qualitative identification of target microorganisms
  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
  • For qualitative identification of Shiga toxin-producing E. coli (STEC)
  • Designed to mimic typical FDA BAM and USDA MLG sample handling procedures
  • Use to report Total STEC or the specific serotype
  • Set of three

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. Receive your final reports within 21 days of the study close date.

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

Each three-sample set contains 3 x 25 g of sterile skim milk powder matrix and three individually packaged lyophilized microorganism pellets. At least one of three of the sample pellets will be positive for STEC (O26, O45, O103, O111, O121, or O145). To use, transfer 25 g of sterile matrix to stomacher bag, add 225 mL of enrichment broth (not included), then add a single sample pellet to the stomacher bag. Process, analyze, then report present or absent for each sample in the set. Three different series available per study.

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

Specifications
  • OrganismShiga toxin-producing E. coli (STEC)
  • Organism typeBacteria
  • Packaging3/PK
  • Product TypeFood Microbiology Proficiency Test
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CMatrix: store at room temperature
  • StrainO26, O45, O103, O111, O121, or O145
  • DescriptionQualitative STEC with Dairy Matrix Proficiency Test Set; 3/PK
Reviews
Q&A