NSI Lab Solutions HPCQC-20X Heterotrophic Plate Count (HPC) Control Standard NCTC 775; 20/PK
Representative image only.
NSI Lab Solutions HPCQC-20X Heterotrophic Plate Count (HPC) Control Standard NCTC 775; 20/PK
NSI Lab Solutions | Item: UX-05002-54
A single pellet contains a pure culture that will yield a specific certified activity level—activity level is stated on the product lot COA
$259.00USD / Pkg of 20
$259.00USD / Pkg of 20
Product Description
Key Features
  • Hydration fluid dissolves culture pellet in less than a minute
  • No dilution needed—hydrate then directly inoculate media
  • Less than 10% RSD lot precision
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • No royalty fee
  • Pack of 20
More About this Item
These microbiological reference materials are designed for use as quantitative control for environmental water analysis. To use, simply transfer the pellet to a sterile room temperature hydration fluid. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Directly inoculate media without serial dilution for convenience.
Use HPC standard as a quantitative control for Heterotrophic/Total Viable counts analyses by Plate count or SimPlate® count method. The Certificate of Analysis (CoA) lists value assignments and acceptance limits for both MPN and plate count methods. Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
NCTC reference cultures obtained from the National Collection of Type Cultures.
SimPlate® is a registered trademark of IDEXX Laboratories, Inc.
Specifications
  • OrganismHPC
  • Packaging20/PK
  • Product TypeEnvironmental Single Cultures
  • Shelf Life Months (months)18
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • StrainNCTC 775
  • DescriptionHeterotrophic Plate Count (HPC) Control Standard NCTC 775; 20/Pk