NSI Lab Solutions PEI-130 Color Wastewater Proficiency Test; 2/PK
Representative image only.
NSI Lab Solutions PEI-130 Color Wastewater Proficiency Test; 2/PK
NSI Lab Solutions | Item: EW-61201-57
Ensure state and accreditation regulation compliance with proficiency testing
$95.00USD / Pkg of 2
$95.00USD / Pkg of 2
Product Description
Key Features
  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • ISO 17043 accredited
  • Four studies per year without order deadlines
  • Supplied in duplicate for convenience and value
  • Two 100-mL whole-volume sample set for determination of Color
  • Formulated in the NELAC range of 10 to 75 CU
More About this Item
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.
These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.
If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.
Specifications
  • Element/IonColor
  • FormLiquid
  • Volume (mL)100
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8 °C
  • Shelf Life (months)24
  • DescriptionColor Wastewater Proficiency Test; 2/PK