NSI Lab Solutions PEO-121 Base/Neutrals Wastewater Proficiency Test; 2/PK

Representative image only.

NSI Lab Solutions PEO-121 Base/Neutrals Wastewater Proficiency Test; 2/PK

NSI Lab Solutions | Item: EW-61202-60

Ensure state and accreditation regulation compliance with proficiency testing

$168.00USD / Pkg of 2

$168.00USD / Pkg of 2
Product Description

Key Features

  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Supplied in duplicate for convenience and value
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • ISO 17043 accredited
  • Two 1.5-mL concentrate set for use with Methods 625/8270

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

The sample design will satisfy PT requirements for any of the following analytes:
1,1-Biphenyl: 30 to 200 µg/L
1,2,4,5-Tetrachlorobenzene: 20 to 200 µg/L
1,2,4-Trichlorobenzene: 20 to 200 µg/L
1,2-Dichlorobenzene: 20 to 200 µg/L
1,2-Diphenylhydrazine: 30 to 200 µg/L
1,3,5-Trinitrobenzene: 20 to 200 µg/L
1,3-Dichlorobenzene: 20 to 200 µg/L
1,3-Dinitrobenzene: 20 to 200 µg/L
1,4-Dichlorobenzene: 20 to 200 µg/L
1,4-Dioxane: 20 to 200 µg/L
1,4-Naphthoquinone: 20 to 200 µg/L
1-Chloronaphthalene: 20 to 200 µg/L
1-Methylnaphthalene: 30 to 200 µg/L
1-Naphthylamine: 20 to 200 µg/L
2,3-Dichloroaniline: 20 to 200 µg/L
2,4-Dinitrotoluene: 20 to 200 µg/L
2,6-Dinitrotoluene: 20 to 200 µg/L
2-Acetylaminofluorene: 20 to 200 µg/L
2-Chloronaphthalene: 20 to 200 µg/L
2-Methylcholanthrene: 10 to 200 µg/L
2-Methylnaphthalene: 20 to 200 µg/L
2-Naphthylamine: 20 to 200 µg/L
2-Nitroaniline: 10 to 200 µg/L
2-Picoline: 20 to 200 µg/L
3,3-Dimethylbenzidine: 20 to 200 µg/L
3,3’-Dichlorobenzidine: 50 to 200 µg/L
3-Methylcholanthrene: 20 to 200 µg/L
3-Nitroaniline: 30 to 200 µg/L
4-Aminobiphenyl: 20 to 200 µg/L
4-Bromophenyl phenyl ether: 20 to 200 µg/L
4-Chloroaniline: 10 to 200 µg/L
4-Chlorophenyl phenyl ether: 20 to 200 µg/L
4-Nitroaniline: 10 to 200 µg/L
4-Nitroquineoline-1-oxide: 20 to 200 µg/L
5-Nitro-o-toluidine: 20 to 200 µg/L
7,12-Dimethylbenz(a)anthracene: 20 to 200 µg/L
a,a-Dimethylphenylamine: 20 to 200 µg/L
Acenaphthene: 10 to 200 µg/L
Acenaphthylene: 10 to 200 µg/L
Acetophenone: 20 to 200 µg/L
Aniline: 30 to 200 µg/L
Anthracene: 10 to 200 µg/L
Atrazine: 30 to 200 µg/L
Benzaldehyde: 30 to 200 µg/L
Benzidine: 200 to 1000 µg/L
Benzo(a)anthracene: 10 to 200 µg/L
Benzo(a)pyrene: 10 to 200 µg/L
Benzo(b)fluoranthene: 20 to 200 µg/L
Benzo(g,h,i)perylene: 10 to 200 µg/L
Benzo(k)fluoranthene: 20 to 200 µg/L
Benzyl alcohol: 30 to 200 µg/L
Benzyl butyl phthalate: 50 to 200 µg/L
bis(2-Chloroethoxy)methane: 20 to 200 µg/L
bis(2-Chloroethyl)ether: 20 to 200 µg/L
2,2’-Oxybis(1-Chloropropane): 30 to 200 µg/L
bis(2-Ethylhexyl)phthalate: 20 to 200 µg/L
Caprolactam: 30 to 200 µg/L
Carbazole: 20 to 200 µg/L
Chlorobenzilate: 20 to 200 µg/L
Chrysene: 10 to 200 µg/L
Di-n-butyl phthalate: 40 to 200 µg/L
Di-n-octyl phthalate: 30 to 200 µg/L
Diallate: 20 to 200 µg/L
Dibenz(a,h)anthracene: 20 to 200 µg/L
Dibenzofuran: 30 to 200 µg/L
Diethyl phthalate: 50 to 200 µg/L
Dimethoate: 20 to 200 µg/L
Dimethyl phthalate: 50 to 200 µg/L
Dinoseb: 20 to 200 µg/L
Diphenyl ether: 20 to 200 µg/L
Disulfoton: 20 to 200 µg/L
Ethyl methanesulfonate : 30 to 200 µg/L
Famphur: 20 to 200 µg/L
Fluoranthene: 30 to 200 µg/L
Fluorene: 10 to 200 µg/L
Hexachlorobenzene: 20 to 200 µg/L
Hexachlorobutadiene: 50 to 200 µg/L
Hexachlorocyclopentadiene: 50 to 200 µg/L
Hexachloroethane: 50 to 200 µg/L
Hexachlorophene: 20 to 200 µg/L
Hexachloropropene: 20 to 200 µg/L
Indeno(1,2,3-c,d)pyrene: 30 to 200 µg/L
Isodrin: 20 to 200 µg/L
Isophorone: 20 to 200 µg/L
Isosafrole: 20 to 200 µg/L
Kepone: 20 to 200 µg/L
m-Dinitrobenzene: 10 to 200 µg/L
Methapyrilene: 20 to 200 µg/L
Methyl methanesulfonate: 10 to 200 µg/L
Methyl parathion: 20 to 200 µg/L
n-Decane: 20 to 200 µg/L
N-Nitroso-di-n-butylamine: 20 to 200 µg/L
N-Nitroso-di-n-propylamine: 30 to 200 µg/L
N-Nitrosodiethylamine: 20 to 200 µg/L
N-Nitrosodimethylamine: 75 to 200 µg/L
N-Nitrosodiphenylamine: 30 to 200 µg/L
N-Nitrosomorpholine: 20 to 200 µg/L
N-Nitrosopiperidine: 20 to 200 µg/L
N-Nitrosopyrrolidine: 20 to 200 µg/L
n-Octadecane: 20 to 200 µg/L
Naphthalene: 20 to 200 µg/L
Nitrobenzene: 20 to 200 µg/L
o,o,o-Triethylphoshorothioate: 20 to 200 µg/L
o-Dinitrobenzene: 10 to 200 µg/L
o-Toluidine: 20 to 200 µg/L
p-Dimethylaminoazobenzene: 20 to 200 µg/L
p-Dinitrobenzene: 10 to 200 µg/L
p-Phenylenediamine: 20 to 200 µg/L
Parathion: 20 to 200 µg/L
Pentachlorobenzene: 20 to 200 µg/L
Pentachlorohexane: 20 to 200 µg/L
Pentachloronitrobenzene: 20 to 200 µg/L
Phenacetin: 20 to 200 µg/L
Phenanthrene: 10 to 200 µg/L
Phorate: 20 to 200 µg/L
Pronamide: 20 to 200 µg/L
Pyrene: 10 to 200 µg/L
Pyridine: 10 to 200 µg/L
Safrole: 20 to 200 µg/L
Sulfotepp: 20 to 200 µg/L
Thionazin: 20 to 200 µg/L

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

Specifications
  • MatrixAcetone/Methanol soution
  • Element/IonMultiple (see the full listing under "More About This Item")
  • FormLiquid
  • Volume (mL)1.5
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8°C
  • UN Number1090/1230
  • Shelf Life (months)24
  • DescriptionBase/Neutrals Wastewater Proficiency Test; 2/PK
Reviews
Q&A