NSI Lab Solutions SQCI-014B Flash Point in Soil PT Express™
Representative image only.
NSI Lab Solutions SQCI-014B Flash Point in Soil PT Express™
NSI Lab Solutions | Item: EW-61202-33
On-demand proficiency test (PT) for when you need results fast
$192.00USD / Each
$192.00USD / Each
Product Description
Key Features
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • One 110 mL sample for Ignitability in the NELAC range of 100 to 200 °F
More About this Item
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.
These environmental chemistry soil and hazardous waste proficiency tests are designed for identification of target chemicals in the soil and underground storage tanks (UST).
Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
Ground Shipping Only.
Specifications
  • Element/IonIgnitability
  • FormLiquid
  • Volume (mL)110
  • Storage SymbolRefrigerate between 2 and 8 °C
  • Shelf Life (months)24
  • DescriptionFlash Point in Soil PT Express™