NSI Lab Solutions SS-29544-6 C. sakazakii Snap-Stick™ Swab; 6/PK
Representative image only.
NSI Lab Solutions SS-29544-6 C. sakazakii Snap-Stick™ Swab; 6/PK
NSI Lab Solutions | Item: UX-05002-01
Easy-to-use Snap-Stick™ culture swab contains a pure microorganism
$190.00USD / Pkg of 6
$190.00USD / Pkg of 6
Product Description
Key Features
  • Hydration fluid dissolves culture pellet in less than a minute
  • Transparent tube ensures visual confirmation of pellet dissolution
  • Removable label prevents sample mix up
  • Glass-free culture swab device ensures user safety
  • No more than two passages from primary culture
  • No royalty fee
  • C. sakazakii NCTC 11467 culture
  • Pack of six
More About this Item
Use these Snap-Stick™ culture tubes containing microorganism pellets for qualitative microbiological analysis. To activate the microorganism, simply apply pressure to the plastic stem valve and release the hydration fluid to dissolve the microorganism pellet. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves in less than a minute with minimum agitation and no incubation. Transparent tube makes it easy to see if the pellet has completely dissolved. Once dissolved, remove the swab from the tube and swipe across the media plate. Each stick comes individually sealed and with a reusable label; use the label to mark your media plate to prevent sample mix up. Store refrigerated.
NCTC reference cultures obtained from the National Collection of Type Cultures.
Specifications
  • OrganismC. sakazakii
  • Packaging6/PK
  • Product TypeSnap-Stick™ Cultures
  • Shelf Life Months (months)24
  • Shipping requirementFrozen ice packs
  • Storage requirementRefrigerate between 2 and 8°C
  • StrainNCTC 11467, 29544
  • DescriptionC. sakazakii Snap-Stick™ Swab; 6/PK