NSI Lab Solutions Microbiological Snap-Stick™ Culture Swabs, A to C
Easy-to-use Snap-Stick™ culture swab contains a pure microorganism
  • Hydration fluid dissolves culture pellet in less than a minute
  • Transparent tube ensures visual confirmation of pellet dissolution
  • Removable label prevents sample mix up
  • Glass-free culture swab device ensures user safety
  • No more than two passages from primary culture
  • No royalty fee
Use these Snap-Stick™ culture tubes containing microorganism pellets for qualitative microbiological analysis. To activate the microorganism, simply apply pressure to the plastic stem valve and release the hydration fluid to dissolve the microorganism pellet. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves in less than a minute with minimum agitation and no incubation. Transparent tube makes it easy to see if the pellet has completely dissolved. Once dissolved, remove the swab from the tube and swipe across the media plate. Each stick comes individually sealed and with a reusable label; use the label to mark your media plate to prevent sample mix up. Store refrigerated.
$68.00 - $190.00USD / Pkg of 2
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Organism Packaging Strain
Availability Pricing
COMPARE
A. flavus 2/PK NRRL 1957
$68.00USD / Pkg of 2
COMPARE
A. flavus 6/PK NRRL 1957
$190.00USD / Pkg of 6
COMPARE
A. fumigatus 2/PK NCTC 982
$68.00USD / Pkg of 2
COMPARE
A. fumigatus 6/PK NCTC 982
$190.00USD / Pkg of 6
COMPARE
A. niger 2/PK NRRL 326, 16888
$68.00USD / Pkg of 2
COMPARE
A. niger 6/PK NRRL 326, 16888
$190.00USD / Pkg of 6
COMPARE
A. terreus 2/PK NCTC 981
$68.00USD / Pkg of 2
COMPARE
A. terreus 6/PK NCTC 981
$190.00USD / Pkg of 6
COMPARE
B. cereus 2/PK NCTC 10320, 11778
$68.00USD / Pkg of 2
COMPARE
B. cereus 6/PK NCTC 10320, 11778
$190.00USD / Pkg of 6
COMPARE
B. subtilis 2/PK NCTC 10400, 6633
$68.00USD / Pkg of 2
COMPARE
B. subtilis 6/PK NCTC 10400, 6633
$190.00USD / Pkg of 6
COMPARE
C. albicans 2/PK NCPF 3179, 10231
$68.00USD / Pkg of 2
COMPARE
C. albicans 6/PK NCPF 3179, 13124
$190.00USD / Pkg of 6
COMPARE
C. sakazakii 2/PK NCTC 11467, 29544
$68.00USD / Pkg of 2
COMPARE
C. sakazakii 6/PK NCTC 11467, 29544
$190.00USD / Pkg of 6
Showing 1- 16 of 16