Reagents Hydrochloric Acid, 37%, ACS Reagent, Poly Bottle, 2.5 L

Representative image only.

Reagents Hydrochloric Acid, 37%, ACS Reagent, Poly Bottle, 2.5 L

Reagents | Mfr: C5101100-2.5A| Item: UX-62286-37

Manufactured using the highest quality raw materials in ISO 9001 facility

$82.30USD / Each

$82.30USD / Each

Buy It Together

Current Item

Reagents Hydrochloric Acid, 37%, ACS Reagent, Poly Bottle, 2.5 L
Reagents Hydrochloric Acid, 37%, ACS Reagent, Poly Bottle, 2.5 L
6228637

$82.30USD / Each
+
Cole-Parmer Hydrochloric Acid, 1.00N, 4 L
8840038

$117.00USD / Each
=
Total Price 199.30 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Minimum Assay 36.5 to 38%
  • Density: 1.18
  • Boiling point: approximately 100 °C (212 °F)
  • Solubility information: miscible
  • pH range: <1
  • 2.5 L poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaHCl
  • ClassAcid
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight36.46
  • PackagingPoly bottle
  • DOT InformationHYDROCHLORIC ACID, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number7647-01-0
  • UN NumberUN1789
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Melting Point C (° C)-28
  • Melting Point F (° F)-18.4
  • DescriptionHydrochloric Acid, 37%, ACS Reagent, Poly Bottle, 2.5 L
Documents & Certificates
Reviews
Q&A