Reagents Isopropanol, Purified, Winpak Container, 20 L

Representative image only.

Reagents Isopropanol, Purified, Winpak Container, 20 L

Reagents | Mfr: C2121500-20E7| Item: EW-62283-80

Manufactured using the highest quality raw materials in ISO 9001 facility

$578.00USD / Each

$578.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless liquid
  • Density: 0.78
  • Flash point: 12 °C (53.6 °F)
  • Refractive index: 1.376 to 1.378nD
  • Solubility information: miscible
  • 20 L Winpak container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaCH3CHOHCH3
  • ClassSolvents
  • ChemistrySolvents
  • Synonyms2-Propanol, Dimethyl carbinol, IPA, Rubbing alcohol
  • FormLiquid
  • Volume (mL)20000
  • Formula Weight60.1
  • Vapor Pressure (torr)31.5026
  • PackagingWinpak container
  • DOT InformationISOPROPANOL, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number67-63-0
  • UN NumberUN1219
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)82.5
  • Boiling Point F (° F)180.5
  • Melting Point C (° C)-90
  • Melting Point F (° F)-130
  • DescriptionIsopropanol, Purified, Winpak Container, 20 L
Documents & Certificates
Reviews
Q&A