Reagents® Isopropanol, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 99%
  • Colorless liquid
  • Density: 0.78
  • Flash point: 12 °C (53.6 °F)
  • Refractive index: 1.376 to 1.378nD
  • Solubility information: miscible
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$57.30 - $626.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Isopropanol, Purified, Winpak Container, 20 L EW-62283-80 Mfr # C2121500-20E7
COMPARE
Solvents Winpak container 20000
$578.00USD / Each
Reagents Isopropanol, Purified, Natural Poly Container, 1 L EW-78911-74 Mfr # C2121500-1A
COMPARE
Solvents Poly container 1000
$139.50USD / Each
Reagents Isopropanol, Purified, Natural Poly Container, 500 mL EW-78911-76 Mfr # C2121500-500A
COMPARE
Solvents Poly container 500
$57.30USD / Each
Reagents Isopropanol, Purified, Natural Poly Container, 4 L EW-78919-86 Mfr # C2121500-4A
COMPARE
Solvents Poly container 4000
$203.75USD / Each
Reagents Isopropanol, Purified, Round Metal Can, 20 L EW-78926-81 Mfr # C2121500-20E6
COMPARE
Solvents Round metal can 20000
$626.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat