Reagents Phosphoric Acid, 2.5L, Purified

Representative image only.

Reagents Phosphoric Acid, 2.5L, Purified

Reagents | Mfr: C5102000-2.5D1| Item: EW-78924-13

Manufactured using the highest quality raw materials in ISO 9001 facility

$220.25USD / Each

$220.25USD / Each
Product Description

Key Features

  • Minimum Assay 85%
  • 7 year warranty

More About this Item

Phosphoric Acid, 500mL, 85% w/w ConcentratIon,Purified Reagent Grade, Colorless, Liquid, 260°C Boiling Point, <2 pH Range, 1.69 Density, 61 Months Shelf Life.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaH3PO4
  • ClassAcid
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight98
  • PackagingClear Glass Bottle
  • DOT InformationPHOSPHORIC ACID SOLUTION, 8
  • UN NumberUN1805
  • CAS Number7664-38-2
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionPhosphoric Acid, 2.5L, Purified
  • Warranty7 year
Documents & Certificates
Reviews
Q&A