Reagents® Phosphoric Acid, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 85%
Phosphoric Acid, 500mL, 85% w/w ConcentratIon,Purified Reagent Grade, Colorless, Liquid, 260°C Boiling Point, <2 pH Range, 1.69 Density, 61 Months Shelf Life.
$69.80 - $220.25USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Phosphoric Acid, 500mL, Purified EW-78916-31 Mfr # C5102000-500C
COMPARE
Acid 500 Phosphoric Acid, 500mL, Purified
$69.80USD / Each
Reagents Phosphoric Acid, 2.5L, Purified EW-78924-13 Mfr # C5102000-2.5D1
COMPARE
Acid 2500 Phosphoric Acid, 2.5L, Purified
$220.25USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat