Reagents Potassium Iodide, ACS Reagent, Poly Pail, 12 kg

Representative image only.

Reagents Potassium Iodide, ACS Reagent, Poly Pail, 12 kg

Reagents | Mfr: C1286000-12KG| Item: EW-78926-64

Manufactured using the highest quality raw materials in ISO 9001 facility

$4,210.00USD / Each

$4,210.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • White solid
  • Density: 3.12
  • Solubility information: 1480 g/L at 25 °C (77 °F)
  • pH range: 6 to 9.2
  • Free flowing
  • 12 kg poly pail
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaKI
  • ClassSalts
  • FormSolid
  • Weight (g)12000
  • Formula Weight166
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7681-11-0
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)1330
  • Boiling Point F (° F)2426
  • Melting Point C (° C)680
  • Melting Point F (° F)1256
  • DescriptionPotassium Iodide, ACS Reagent, Poly Pail, 12 kg
  • Warranty7 year
Documents & Certificates
Reviews
Q&A