Reagents® Potassium Iodide, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 99%
  • White solid
  • Density: 3.12
  • Solubility information: 1480 g/L at 25 °C (77 °F)
  • pH range: 6 to 9.2
  • Free flowing
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$50.20 - $16,610.00USD / Each

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Packaging Weight (g) Description
Availability Pricing
Reagents Potassium Iodide, ACS Reagent, Fiber Drum, 100 lb EW-62282-51 Mfr # C1286000-100LB
COMPARE
Fiber drum 4536 Potassium Iodide, ACS Reagent, Fiber Drum, 100 lb
$16,610.00USD / Each
Reagents Potassium Iodide, ACS Reagent, Poly Pail, 10 kg EW-62282-52 Mfr # C1286000-10KG
COMPARE
Poly pail 10000 Potassium Iodide, ACS Reagent, Poly Pail, 10 kg
$4,145.00USD / Each
Reagents Potassium Iodide, ACS Reagent, Natural Poly Container, 2.25 kg EW-62282-53 Mfr # C1286000-2.5A5
COMPARE
Poly container 2250 Potassium Iodide, ACS Reagent, Natural Poly Container, 2.25 kg
$1,625.00USD / Each
Reagents Potassium Iodide, ACS Reagent, Natural Poly Container, 3 kg EW-62282-54 Mfr # C1286000-3A5
COMPARE
Poly container 3000 Potassium Iodide, ACS Reagent, Natural Poly Container, 3 kg
$1,910.00USD / Each
Reagents Potassium Iodide, ACS Reagent, Wide-Mouth Amber Glass Bottle, 50 g EW-62282-55 Mfr # C1286000-50C2
COMPARE
Wide-mouth amber glass bottle 50 Potassium Iodide, ACS Reagent, Wide-Mouth Amber Glass Bottle, 50 g
$50.20USD / Each
Reagents Potassium Iodide, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g EW-78913-14 Mfr # C1286000-100C2
COMPARE
Wide-mouth amber glass bottle 100 Potassium Iodide, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g
$169.25USD / Each
Reagents Potassium Iodide, ACS Reagent, Wide-Mouth Container, 500 g EW-78913-16 Mfr # C1286000-500A5
COMPARE
Wide-mouth container 500 Potassium Iodide, ACS Reagent, Wide-Mouth Container, 500 g
$460.00USD / Each
Reagents Potassium Iodide, ACS Reagent, Poly Pail, 12 kg EW-78926-64 Mfr # C1286000-12KG
COMPARE
Poly pail 12000 Potassium Iodide, ACS Reagent, Poly Pail, 12 kg
$4,210.00USD / Each
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat