Reagents Salicylic Acid, 100g, ACS Reagent

Representative image only.

Reagents Salicylic Acid, 100g, ACS Reagent

Reagents | Mfr: C1113000-100C2| Item: EW-78913-44

Manufactured using the highest quality raw materials in ISO 9001 facility

$55.40USD / Each

$55.40USD / Each
Product Description

Key Features

  • Minimum Assay 99%, Crystal
  • 7 year warranty

More About this Item

Salicylic Acid, 100g, ACS Reagent Grade, 69-72-7 CAS Number, C7H6O3 Chemical Formula, 138.12g/mol Molecular Weight, Colorless/White, Solid, 211°C Boiling Point, 159°C Melting Point, 157°C Flash Point, 1.44 Density, 60 Months Shelf Life.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC7H6O3
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight138.12
  • Vapor Pressure (torr)0.2325
  • PackagingAmber Jar
  • CAS Number69-72-7
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionSalicylic Acid, 100g, ACS Reagent
  • Warranty7 year
Documents & Certificates
Reviews
Q&A