Reagents® Salicylic Acid, ACS Reagent
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 99%, Crystal
Salicylic Acid, 100g, ACS Reagent Grade, 69-72-7 CAS Number, C7H6O3 Chemical Formula, 138.12g/mol Molecular Weight, Colorless/White, Solid, 211°C Boiling Point, 159°C Melting Point, 157°C Flash Point, 1.44 Density, 60 Months Shelf Life.
$55.40 - $130.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Weight (g) Description
Availability Pricing
Reagents Salicylic Acid, 100g, ACS Reagent EW-78913-44 Mfr # C1113000-100C2
COMPARE
Salts 100 Salicylic Acid, 100g, ACS Reagent
$55.40USD / Each
Reagents Salicylic Acid, 500g, ACS Reagent EW-78913-45 Mfr # C1113000-500A5
COMPARE
Salts 500 Salicylic Acid, 500g, ACS Reagent
$130.00USD / Each