Reagents Sodium Metabisulfite, Purified, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Sodium Metabisulfite, Purified, Natural Poly Container, 2.5 kg

Reagents | Mfr: C2320100-2.5A5| Item: EW-78919-40

Manufactured using the highest quality raw materials in ISO 9001 facility

$97.10USD / Each

$97.10USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.48
  • Melting point: 150 °C (302 °F), decomposes
  • Solubility information: 667 g/L at 25 °C (77 °F)
  • 2.5 kg natural poly container
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

5 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaNa2O5S2
  • ClassSalts
  • FormSolid
  • Weight (g)2500
  • Formula Weight190.11
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7681-57-4
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Metabisulfite, Purified, Natural Poly Container, 2.5 kg
  • Warranty5 years
Documents & Certificates
Reviews
Q&A