Reagents® Sodium Metabisulfite, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless/white solid
  • Density: 1.48
  • Melting point: 150 °C (302 °F), decomposes
  • Solubility information: 667 g/L at 25 °C (77 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$43.90 - $472.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Weight (g) Description
Availability Pricing
Reagents Sodium Metabisulfite, Purified, Fiber Drum, 50 lb EW-62285-85 Mfr # C2320100-50F5
COMPARE
Salts 22680 Sodium Metabisulfite, Purified, Fiber Drum, 50 lb
$472.00USD / Each
Reagents Sodium Metabisulfite, Purified, Wide-Mouth Container, 500 g EW-78913-94 Mfr # C2320100-500A5
COMPARE
Salts 500 Sodium Metabisulfite, Purified, Wide-Mouth Container, 500 g
$43.90USD / Each
Reagents Sodium Metabisulfite, Purified, Natural Poly Container, 2.5 kg EW-78919-40 Mfr # C2320100-2.5A5
COMPARE
Salts 2500 Sodium Metabisulfite, Purified, Natural Poly Container, 2.5 kg
$97.10USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat