Thermo Scientific Nunc 267427 Microplates with Lid, 96-Well, Polystyrene, Sterile, Non-Treated; 50/Cs

Representative image only.

Thermo Scientific Nunc 267427 Microplates with Lid, 96-Well, Polystyrene, Sterile, Non-Treated; 50/Cs

Thermo Scientific | Mfr: 267427| Item: EW-01930-66

Minimize plate evaporation and the risks associated with edge effect

$443.00USD / Case of 50

$443.00USD / Case of 50
Product Description

Key Features

  • Excellent optical performance
  • Reduced edge effects
  • Use 38% more of your plate – cells are still viable
  • Non-treated surface is available for suspension cell culture or general solution-based assays

More About this Item

The Edge™ 2.0 96-well plates provide an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect. The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL of sterile water, buffer, or media. When filled with fluid, the surrounding moat acts as an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste. Choose from Nunclon™ treated or non-treated surfaces.

Specifications
  • Number of Wells96
  • Well Volume (µL)400
  • Well Bottom Diameter (mm)6.7
  • Well Top Diameter (mm)7.1
  • Well ShapeFlat
  • Surface TreatmentNon-Treated
  • Lid (Included)Yes
  • SterileYes
  • Plate ColorClear
  • MaterialPolystyrene
  • Width (mm)86
  • Length (mm)128
  • DescriptionMicroplates with Lid, 96-Well, Polystyrene, Sterile, Non-Treated; 50/Cs
Reviews
Q&A