Thermo Scientific Precision GPSSL05 Stainless Steel Cover, 2S/5L
Representative image only.
Thermo Scientific Precision GPSSL05 Stainless Steel Cover, 2S/5L
Thermo Scientific | Mfr: GPSSL05| Item: EW-03710-76
Gable cover ensures temperatures are maintained in water baths
$520.00USD / Each
$520.00USD / Each
Product Description
More About this Item
These stainless steel gabled bath covers are ideal for use with the 2, 5, 10 and 28 L Thermo Scientific Precision general-purpose digital water baths.
Specifications
  • DescriptionStainless Steel Cover, 2S/5L