Thermo Scientific Precision Stainless Steel Bath Covers
Gable cover ensures temperatures are maintained in water baths
These stainless steel gabled bath covers are ideal for use with the 2, 5, 10 and 28 L Thermo Scientific Precision general-purpose digital water baths.
$520.00 - $565.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific Precision GPSSL05 Stainless Steel Cover, 2S/5L EW-03710-76 Mfr # GPSSL05
COMPARE
Stainless Steel Cover, 2S/5L
$520.00USD / Each
Thermo Scientific Precision GPSSL10 Stainless Steel Cover, 10L EW-03710-77 Mfr # GPSSL10
COMPARE
Stainless Steel Cover, 10L
$525.00USD / Each
Thermo Scientific Precision GPSSL20 Stainless Steel Gable Cover, 20/28L EW-03710-78 Mfr # GPSSL20
COMPARE
Stainless Steel Gable Cover, 20/28L
$565.00USD / Each