Vector Laboratories Vector NovaRed Substrate Kit, Red, 300 ml

Representative image only.

Vector Laboratories Vector NovaRed Substrate Kit, Red, 300 ml

Vector Laboratories | Mfr: SK-4800| Item: EW-93952-00

Successfully label multiple antigens in same tissue section

$299.75USD / Each

$299.75USD / Each
Product Description

Key Features

  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
  • 1 year warranty

More About this Item

These enzyme substrate kits for use with peroxidase detection systems offer a wide variety of applications. Numerous color choices are available for staining antigens in pigmented tissues and melanomas, as well as for localizing multiple antigens in the same tissue section. Viewing options for the substrates include brightfield, fluorescence, darkfield, electron microscopy, and spectral imaging.

The substrates also come in a varying degree of sensitivities. The ImmPACT peroxidase substrates are about 2-4 times more sensitive than the original peroxidase substrate kits.

Vector NovaRed and ImmPACT NovaRed Substrate Kits produce red reaction products. Both substrate kits are useful as alternatives to DAB or as a second color for multiple antigen labeling.

1 Year Warranty
Specifications
  • ColorRed
  • Volume (mL)300
  • DescriptionSubstrate Kit, Red, 300 ml
  • Warranty1 year
Reviews
Q&A