Atago Special-Scale Digital Pocket Refractometers
Measure concentration of specific liquids with these special-scale refractometers
These compact refractometers give you a direct reading scale of the concentration of some common liquids. Rugged IP65-rated design is ideal for indoor and field use. Simply apply two to three drops of sample onto the prism, press the start key, and the value is displayed in seconds. Digital display eliminates reading errors common with handheld refractometers. Cleanup is simple — easily clean under running water.
$656.80 - $784.35USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Resolution Description
Availability Pricing
Atago PAL-03S Special-scale Refractometer, Sodium Chloride EW-02941-01 Mfr # 4403 (PAL-03S)
COMPARE
±0.2% (NaCl) 0.1% (Brix) Special-scale Refractometer, Sodium Chloride
$784.35USD / Each
Atago PAL-06S Special-scale Refractometer, Salinity of Seawater EW-02941-02 Mfr # 4406 (PAL-06S)
COMPARE
±2.0% (salinity) 1% (salinity) Special-scale Refractometer, Salinity of Seawater
$745.50USD / Each
Atago PAL-39S Special-scale Refractometer, Hydrogen Peroxide EW-02941-06 Mfr # 4439 (PAL-39S)
COMPARE
±0.6% (HOOH) 0.2% (HOOH) Special-scale Refractometer, Hydrogen Peroxide
$750.25USD / Each
Atago PAL-40S Special-scale Refractometer, Sodium Hydroxide EW-02941-07 Mfr # 4440 (PAL-40S)
COMPARE
±0.2% (NaOH) 0.1% (NaOH) Special-scale Refractometer, Sodium Hydroxide
$656.80USD / Each
Atago PAL-UREA Digital Pocket Urea Refractometer; 0.0 to 53% EW-02941-13 Mfr # 4518 (PAL-UREA)
COMPARE
±0.3% (urea) 0.1% (urea) Digital Pocket Urea Refractometer; 0.0 to 53%
$776.50USD / Each