Cole-Parmer® Cleanroom Male Luer Locking Plugs, Straight Adapter
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Seal luer ports with a secure and leak-proof connection
  • Luer fittings allow for a simple yet secure twist-lock connection
  • Ideal for capping off fluid transfer system
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
These Cole-Parmer® cleanroom fittings are high-quality PVC-free fittings, ideal in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a female luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
$7.30 - $13.20USD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Connection Type-Port 1 Material Description
Availability Pricing
COMPARE
Luer to Plug Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Rotating Male Luer Lock; 10/Pk
$9.20USD / Pkg of 10
COMPARE
Luer to Plug Fitting, Straight Adapter, White Nylon, Cleanroom Packed, Male Luer Lock; 10/Pk
$11.50USD / Pkg of 10
COMPARE
Luer to Plug Fitting, Straight Adapter, Natural Kynar®, Cleanroom Packed, Male Luer Lock; 10/Pk
$13.20USD / Pkg of 10
COMPARE
Luer to Plug Fitting, Straight Adapter, Natural Nylon, Cleanroom Packed, Male Luer Lock; 10/Pk
$7.30USD / Pkg of 10
COMPARE
Luer to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, Cleanroom Packed, Male Luer Lock; 10/Pk
$7.60USD / Pkg of 10