Cole-Parmer® Cleanroom Male Luer Rotating Plugs, Straight Adapter
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a female luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
$7.30 - $13.20USD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Luer Connection Type (Port 1) Description
Availability Pricing
COMPARE
Polycarbonate Rotating Male Luer Luer to Plug Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Rotating Male Luer; 10/Pk
$9.20USD / Pkg of 10
COMPARE
Nylon Rotating Male Luer Luer to Plug Fitting, Straight Adapter, White Nylon, Cleanroom Packed, Rotating Male Luer; 10/Pk
$11.50USD / Pkg of 10
COMPARE
PVDF Rotating Male Luer Luer to Plug Fitting, Straight Adapter, Natural Kynar®, Cleanroom Packed, Rotating Male Luer; 10/Pk
$13.20USD / Pkg of 10
COMPARE
Nylon Rotating Male Luer Luer to Plug Fitting, Straight Adapter, Natural Nylon, Cleanroom Packed, Rotating Male Luer; 10/Pk
$7.30USD / Pkg of 10
COMPARE
Polypropylene Rotating Male Luer Luer to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, Cleanroom Packed, Rotating Male Luer; 10/Pk
$7.60USD / Pkg of 10