Cole-Parmer® Fluorimeter Cuvettes (Glass and Quartz) with Cover of PTFE Resin
Select a pathlength to suit your analytical needs
  • Standard size cuvettes are the type most often used for routine analytical needs (83301-02 to 83301-07)
  • Use skus 83303-02 to 83303-17 for fluorimeters
Choose from glass or quartz cuvettes to meet your application needs.
$98.60 - $492.75USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
COMPARE
Glass 1.7 5
$98.60USD / Each
COMPARE
Glass 3.5 10
$103.50USD / Each
COMPARE
Glass 7 20
$131.25USD / Each
COMPARE
Glass 10.5 40
$142.50USD / Each
COMPARE
Glass 14 50
$189.25USD / Each
COMPARE
Quartz 3.5 10
$283.25USD / Each
COMPARE
Quartz 7 20
$391.00USD / Each
COMPARE
Quartz 10.5 40
$492.75USD / Each
COMPARE
Quartz 14 50
$316.25USD / Each
Showing 1- 9 of 9