Crosstex Self-Seal Sterilization Pouches
Provides breathability, a sterile barrier, and indication of processing without the need to heat seal
  • Perfect for small instruments that require frequent sterilization
  • Multi-parameter pouches confirm all sterilization criteria — time, temperature, and steam
Standard Pouches feature triple seal edges — three separate heat sealed rails secure edges from separation or tearing by instruments. Constructed of wet strength treated, virgin medical grade paper that meets or exceeds medical pouch standards. Blue tinted transparent film facilitates easy viewing of instruments and identification of punctures or tears in film. Easily visible external process indicator arrows change color with sterilization heat. Follow CDC recommendations and insert internal indicators inside puches to confirm sterility inside (sold separately).
Multi-Parameter Pouches provide the highest level of sterility assurance. Eliminate the extra step of adding separate indicators to each pouch with these multi-parameter indicators that meet all criteria for sterilization including time, temperature, and presence of steam with internal and external indicators printed directly on the pouch. Meets ISO and CDC standards to ensure effective sterilization.
$249.80 - $404.00USD / Case of 4000
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Width (in) Description
Availability Pricing
COMPARE
2 8 Self-Seal Sterilization Pouches, Standard, 2" X 8"; 200/Box, 4,000/Case
$325.00USD / Case of 4000
Crosstex Self-Seal Sterilization Pouches, Standard, 2.25" X 4"; 200/Box, 4,000/Case EW-10703-38 Mfr # SCXX
COMPARE
2 1/4 4 Self-Seal Sterilization Pouches, Standard, 2.25" X 4"; 200/Box, 4,000/Case
$256.00USD / Case of 4000
COMPARE
2 3/4 9 Self-Seal Sterilization Pouches, Standard, 2.75" X 9"; 200/Box, 4,000/Case
$337.00USD / Case of 4000
COMPARE
3 1/4 9 Self-Seal Sterilization Pouches, Standard, 3.25" X 9"; 200/Box, 4,000/Case
$367.00USD / Case of 4000
COMPARE
3 1/2 9 Self-Seal Sterilization Pouches, Standard, 3.5" X 9"; 200/Box, 4,000/Case
$341.00USD / Case of 4000
Crosstex Self-Seal Sterilization Pouches, Standard, 3.5" X 9";  500/Box, 5,000/Case EW-10703-43 Mfr # SCS5
COMPARE
3 1/2 9 Self-Seal Sterilization Pouches, Standard, 3.5" X 9"; 500/Box, 5,000/Case
$356.50USD / Case of 5000
COMPARE
7 1/2 13 Self-Seal Sterilization Pouches, Standard, 7.5" X 13"; 200/Box, 1,000/Case
$249.80USD / Case of 1000
Crosstex Self-Seal Sterilization Pouches, Standard, 12" X 15"; 100/Box, 500/Case EW-10703-48 Mfr # SCL1215
COMPARE
12 15 Self-Seal Sterilization Pouches, Standard, 12" X 15"; 100/Box, 500/Case
$280.30USD / Case of 500
Crosstex Self-Seal Sterilization Pouches, Standard, 12" x 18"; 100/Box, 500/Case EW-10703-49 Mfr # SCL1218
COMPARE
12 18 Self-Seal Sterilization Pouches, Standard, 12" x 18"; 100/Box, 500/Case
$309.50USD / Case of 500
Crosstex Self-Seal Sterilization Pouches, Multiparameter, 2.25" X 4"; 200/Box, 4,000/Case EW-10703-51 Mfr # SCXX2
COMPARE
2 1/4 4 Self-Seal Sterilization Pouches, Multiparameter, 2.25" X 4"; 200/Box, 4,000/Case
$329.00USD / Case of 4000
Crosstex Self-Seal Sterilization Pouches, Multiparameter, 3.5" X 9"; 200/Box, 4,000/Case EW-10703-52 Mfr # SCS2
COMPARE
3 1/2 9 Self-Seal Sterilization Pouches, Multiparameter, 3.5" X 9"; 200/Box, 4,000/Case
$404.00USD / Case of 4000
Crosstex Self-Seal Sterilization Pouches, Multiparameter, 5.25" X 10"; 200/Box, 2,000/Case EW-10703-53 Mfr # SCM2
COMPARE
5 1/4 10 Self-Seal Sterilization Pouches, Multiparameter, 5.25" X 10"; 200/Box, 2,000/Case
$289.50USD / Case of 2000
Crosstex Self-Seal Sterilization Pouches, Multiparameter, 12" X 18"; 100/Box, 500/Case EW-10703-57 Mfr # SCL12182
COMPARE
12 18 Self-Seal Sterilization Pouches, Multiparameter, 12" X 18"; 100/Box, 500/Case
$350.50USD / Case of 500
Showing 1- 13 of 13