Crystal XP2i Digital Pressure Gauges with Data Logging
Collect up to 6,000 data points — Ideal for oil and gas industry
  • Change data collection parameters from either a computer or the keypad of an XP2i
  • Start and stop multiple collection runs from the XP2i keypad
  • Switch an XP2i from standard to DataLogger mode in minutes
  • With the DataLoggerXP option, this gauge is a unique, high accuracy, intrinsically safe data logging solution
The DataLogger XP software and an XP2i digital test gauge form an integrated data logging solution. While the XP2i performs all of the actual data collection, DataLoggerXP handles the transfer and saving of that data to a computer’s hard drive. Once measurements have been collected and transferred, DataLoggerXP can save the data files directly into a spreadsheet.
Available in the USA only.
$2,903.00 - $3,010.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Dial Size (in) Max Pressure (PSI)
Availability Pricing
Crystal 10KPSIXP2I-DL Digital Pressure Gauge with Data Logger, 10,000 psi, Xp Software EW-68436-90 Mfr # 10KPSIXP2I-DL
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 10
$3,010.00USD / Each
Crystal 15PSIXP2I-DL Digital Pressure Gauge with Data Logger, 15 psi, Xp Software EW-68436-91 Mfr # 15PSIXP2I-DL
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 15
$3,010.00USD / Each
Crystal 1KPSIXP2I-DL Digital Pressure Gauge with Data Logger, 1000 psi, Xp Software EW-68436-92 Mfr # 1KPSIXP2I-DL
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 1000
$2,903.00USD / Each
Crystal 300PSIXP2I-DL Digital Pressure Gauge with Data Logger, 300 psi, Xp Software EW-68436-93 Mfr # 300PSIXP2I-DL
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 300
$2,903.00USD / Each
Crystal 2000PSIXP2I-DL Digital Pressure Gauge with Data Logger, 2000 psi,  Xp Software EW-68436-94 Mfr # 2KPSIXP2I-DL
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 2000
$2,903.00USD / Each
Crystal 3KPSIXP2I-DL Digital Pressure Gauge with Data Logger, 3000 psi, Xp Software EW-68436-96 Mfr # 3KPSIXP2I-DL
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 3000
$2,903.00USD / Each
Crystal 5KPSIXP2I-DL Digital Pressure Gauge with Data Logger, 5000 psi, Xp Software EW-68436-98 Mfr # 5KPSIXP2I-DL
COMPARE
±0.1% of reading (20% to 100% of full scale); ±0.02% full-scale (below 20% of full scale) 4.5 5000
$2,903.00USD / Each
Showing 1- 7 of 7