CYTIVA Whatman™ 50 mm In-Line Filters
Versatile and cost-effective filters for large-volume samples
  • PTFE membrane filters are ideal for chemically aggressive solutions. Use as protective vents or for in-line filtration and isolation applications. Filter 29700-76 features a polypropylene prefilter
  • PES aqueous solution filters are low protein binding and contain no surfactants. A glass microfiber prefilter extends the life of the membrane
  • Polypropylene filters have excellent flow rate characteristics and are ideal for filtering up to 1 L of aqueous and solvent samples
Filter up to one liter with one device; filtration area is 16 cm2. Filter devices can be used in conjunction with a syringe or connected in-line (6 to 10 mm hose) via stepped hose barbs. Autoclavable, a high-purity polypropylene housing maintains sample purity. Hold up volume with air purge is less than 0.1 mL. Maximum operating pressure is 60 psi (4.1 bar).
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
$268.30 - $351.75USD / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Material Pore Size (µm)
Availability Pricing
Whatman 6720-5001 50 mm In-Line Filters, PTFE, 0.1µm, 10/pk EW-29700-70 Mfr # 6720-5001
COMPARE
PTFE 0.1
$268.30USD / Pkg of 10
Whatman 6720-5002 50 mm In-Line Filters, PTFE, 0.2µm, 10/pk EW-29700-72 Mfr # 6720-5002
COMPARE
PTFE 0.2
$304.25USD / Pkg of 10
Whatman 6720-5045 50 mm In-Line Filters, PTFE, 0.45µm, 10/pk EW-29700-74 Mfr # 6720-5045
COMPARE
PTFE 0.45
$301.65USD / Pkg of 10
Whatman 6721-5010 50 mm In-Line Filters, PTFE, 1.0µm, 10/pk EW-29700-76 Mfr # 6721-5010
COMPARE
PTFE 1
$291.65USD / Pkg of 10
Whatman 6724-5002 Sterile Polydisc AS, 0.2 um PES w/GMF Prefilter, 50 mm Dia; 10/Pk EW-29700-78 Mfr # 6724-5002
COMPARE
Polyethersufone (PES) 0.2
$336.55USD / Pkg of 10
Whatman 6724-5045 Sterile Polydisc AS, 0.45 um PES w/GMF Prefilter, 50 mm Dia; 10/Pk EW-29700-80 Mfr # 6724-5045
COMPARE
Polyethersufone (PES) 0.45
$351.75USD / Pkg of 10
Whatman 6728-5050 50 mm In-Line Filters, Polypropylene, 5.0µm, 10/pk EW-29700-86 Mfr # 6728-5050
COMPARE
Polypropylene 5
$289.55USD / Pkg of 10
Whatman 6728-5100 50 mm In-Line Filters, Polypropylene, 10.0µm, 10/pk EW-29700-88 Mfr # 6728-5100
COMPARE
Polypropylene 10
$281.95USD / Pkg of 10
Showing 1- 8 of 8