Diba Omnifit® Flared Tubing Assemblies with Click-N-Seal® 1/4"-28 UNF(M) Fittings, 1/32"ID x 1/16"OD

Flared tubing and fitting assemblies offer fast leak-free connections
  • When the cap clicks, you know the seal is tight
  • Flared tubing sits tightly against a flat bottom of the port
  • Only the tube material is in the fluid path
  • Available with polycarbonate (PC), acetal, and PEEK fittings
Self-torquing Click-N-Seal fittings create a seal without tools. As the fitting is tightened the resistance of the cap increases until the correct torque is reached. Acetal and PEEK fittings' compact design easily fits in tight spaces.
$33.40 - $43.70USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Tubing Formulation Description
Availability Pricing
Diba Click-N-Seal™ Plus Tubing Assembly, 1/4"-28 UNF(M) with flare, FEP/Acetal, 1/16" OD x 24" L; 1/ea EW-21943-44 Mfr # DB200-15047
COMPARE
2 FEP Click-N-Seal™ Plus Tubing Assembly, 1/4"-28 UNF(M) with flare, FEP/Acetal, 1/16" OD x 24" L; 1/ea
$43.70USD / Each
Diba Click-N-Seal™ Plus Tubing Assembly, 1/4"-28 UNF(M) with flare, PTFE/Acetal, 1/16" OD x 24" L; 1/ea EW-21943-46 Mfr # DB200-15049
COMPARE
2 PTFE Click-N-Seal™ Plus Tubing Assembly, 1/4"-28 UNF(M) with flare, PTFE/Acetal, 1/16" OD x 24" L; 1/ea
$34.00USD / Each
Diba Click-N-Seal™ Tubing Assembly, 1/4"-28 UNF(M) with flare, PTFE/PC, 1/16" OD x 24" L; 1/ea EW-21943-49 Mfr # DB200-15052
COMPARE
2 PTFE Click-N-Seal™ Tubing Assembly, 1/4"-28 UNF(M) with flare, PTFE/PC, 1/16" OD x 24" L; 1/ea
$33.40USD / Each
Diba Click-N-Seal™ Ultra Tubing Assembly, 1/4"-28 UNF(M) with flare, PTFE/PEEK, 1/16" OD x 24" L; 1/ea EW-21943-51 Mfr # DB200-15054
COMPARE
2 PTFE Click-N-Seal™ Ultra Tubing Assembly, 1/4"-28 UNF(M) with flare, PTFE/PEEK, 1/16" OD x 24" L; 1/ea
$43.70USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat