DuPont™ Liveo™ Pharma-50 Platinum-Cured Silicone Tubing
Ideal for transfer of high-purity fluids in pharmaceutical and biotechnological applications
  • Provides excellent flexibility
  • Offers low extractables
  • Contains no peroxide by-products, chlorophenyls or PCBs
  • Free from organic plasticizers, phthalates or latex additives
  • Easily sterilized
  • Meets USP Class VI, FDA registered and ISO 9001:2015 requirements
The translucent, platinum-cured silicone DuPont Liveo Pharma-50 tubing is designed to transfer ultra-pure liquids in pharmaceutical and biotechnological processes where contamination is a concern. The tubing is stable over a wide temperature range, is highly resilient, impairs no taste or odor, and offers a non-wetting surface.
Made from BioMedical Grade elastomer, the tubing exceeds United States Pharmacopeia (USP®) Class VI Plastics Test Requirements. It also meets the European Pharmacopoeia monograph 3.1.9. “Silicone elastomer for closures and tubing.” The tubing is easily sterilized by ethylene oxide, autoclave steam for 30 minutes at 120 °C, and gamma radiation at 50 kGy.
ITEM(S) ON SALE
$128.00 - $561.00USD / Pkg of 1
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 1/8" ID x 1/4" OD; 50 Ft EW-50121-85 Mfr # 3100430
COMPARE
1/8 1/4 1/16
$213.25USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 3/16" ID x 3/8" OD; 50 Ft EW-50121-86 Mfr # 3100456
COMPARE
3/16 3/8 3/32
$287.75USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 3/16" ID x 5/16" OD; 50 Ft EW-50121-87 Mfr # 3100448
COMPARE
3/16 5/16 1/16
$191.75USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 1/4" ID x 1/2" OD; 50 Ft EW-50121-88 Mfr # 3100472
COMPARE
1/4 1/2 1/8
$386.00USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 1/4" ID x 3/8" OD; 50 Ft EW-50121-89 Mfr # 4006922
COMPARE
1/4 3/8 1/16
$248.00USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 1/4" ID x 7/16" OD; 50 Ft EW-50121-90 Mfr # 3100464
COMPARE
1/4 7/16 3/32
$146.00USD / Pkg of 1
$311.50USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 3/8" ID x 1/2" OD; 50 Ft EW-50121-91 Mfr # 4006923
COMPARE
3/8 1/2 1/16
$128.00USD / Pkg of 1
$274.25USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 3/8" ID x 5/8" OD; 50 Ft
Always in Stock
EW-50121-92 Mfr # 3100499
COMPARE
3/8 5/8 1/8
$502.00USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 3/8" ID x 9/16" OD; 50 Ft EW-50121-93 Mfr # 3100481
COMPARE
3/8 9/16 3/32
$385.00USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 1/2" ID x 11/16" OD; 50 Ft EW-50121-94 Mfr # 4006925
COMPARE
1/2 11/16 3/32
$203.00USD / Pkg of 1
$435.00USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 1/2" ID x 3/4" OD; 50 Ft EW-50121-95 Mfr # 3100502
COMPARE
1/2 3/4 1/8
$561.00USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 3/4" ID x 1" OD; 50 Ft EW-50121-96 Mfr # 3100511
COMPARE
3/4 1 1/8
$468.00USD / Pkg of 1
$991.00USD / Pkg of 1
DuPont Liveo Pharma 50 Platinum-Cured Silicone Tubing, 5/8" ID x 7/8" OD; 50 Ft EW-50121-97 Mfr # 3318184
COMPARE
5/8 7/8 1/8
$360.00USD / Pkg of 1
$761.00USD / Pkg of 1
Showing 1- 13 of 13