Environmental Express® Disposable Dispensing Funnels
Convenience of disposable, unbreakable funnels for oil and grease analysis
  • Choose between UltraPrep and UltraFlow extraction funnels
  • Available in 47-mm; Pack of 20
StepSaver streamlines the extraction process by allowing the analyst to collect the solvent with eluted oil and grease directly into a collection flask. As fluid enters the collection flask, it passes through a drying cartridge to remove excess water. It is ready to be evaporated without a separate drying step.
$191.75 - $245.85USD / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
UltraFlow® Funnel with 47 mm Filters, for the StepSaver; 20/PK
$191.75USD / Pkg of 20
COMPARE
UltraFlow® Funnel with 47 mm Filters, for the J.T. Baker® SPE apparatus; 20/PK
$221.95USD / Pkg of 20
COMPARE
UltraPrep® Funnel with 47 mm Filters, for the StepSaver Oil and Grease Extraction System; 20/PK
$216.40USD / Pkg of 20
COMPARE
UltraPrep® Funnel with 47 mm Filters, for the J.T. Baker® Oil and Grease Extraction Apparatus; 20/PK
$245.85USD / Pkg of 20