Environmental Express® HotBlock™ 200 Digestion Controllers
Control one or two block heaters
  • Choose your controller, then block (or blocks) to suit your metals digestion needs
  • Mount controllers outside the fume hood on metal surfaces, in an available outlet, or place on a benchtop
  • Same dependability with increased temperature range
Blocks and controllers are sold separately.
Perform a wider range of experiments and digestions with the expanded temperature range—up to 200°C* with the new HotBlock 200! Controllers feature automatic shut-off, alarms, and ability to save profiles with ramp and soak settings. The dual-block controller can control up to two blocks independently and simultaneously. Safely place the external controller outside the fume hood. Controllers may be mounted to a metal surface using the magnets, or on an electrical outlet.
$1,442.60 - $2,161.65USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Blocks Power (VAC)
Availability Pricing
COMPARE
1 120
$1,442.60USD / Each
COMPARE
2 120
$2,161.65USD / Each