Extech EX350 Series True RMS Multimeters with Noncontact Voltage Detection
Quickly check for live wires without touching potential voltage sources
  • LoZ low impedance mode reduces false readings from ghost voltages
  • Low pass filter reduces noise by attenuating high frequencies for more accurate readings
  • Data hold, minimum, maximum and max-min functions capture important data
  • Relative mode used to zero capacitance and adjust offset
  • Auto power-off conserves battery
  • Choose model with or without thermocouple temperature measurement
Increase safety of technicians with noncontact voltage detection and LoZ mode. True RMS and low pass filter provide accurate readings of variable frequency signals.
$100.00 - $129.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) Current (AC/DC) True Rms
Availability Pricing
Extech EX350 TrueRMS Multimeter w/ Noncontact Voltage Detection EW-20027-01 Mfr # EX350
COMPARE
600 0.1 µA to 10.00 A Yes
$100.00USD / Each
Extech EX355 TrueRMS Multimeter w/ Noncontact Voltage Detection and Temperature EW-20027-03 Mfr # EX355
COMPARE
600 0.1 µA to 10.00 A Yes
$129.00USD / Each