GFG G450 Portable Multigas Monitors
Compact, economical, and reliable
  • AutoCal® automatic calibration Extra large, invertible, heterochromatic display Rotating 360° bright, flashing alarms Loud 103 dB buzzer Rugged design, IP67 (dust and watertight) 24 hours of continuous operation Only 10 ounces plus integral rubberized tough housing
This compact and lightweight gas detector continuously measures gas concentration in ambient air. It's functional and watertight design (IP67) incorporates an impact-proof, rubberized housing to meet the toughest requirements of harsh environments.
$1,435.00 - $1,630.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Gases Measured
Availability Pricing
GFG Instrumentation G450 Portable Gas Monitor; O2, CO, Combustibles EW-10107-80 Mfr # G450-11420
COMPARE
CO, combustibles, O2
$1,435.00USD / Each
GFG Instrumentation G450 Portable Gas Monitor; O2, H2S, CO, Combustibles EW-10107-81 Mfr # G450-11440
COMPARE
CO, combustibles, H2S, O2
$1,630.00USD / Each