ICI Quick Ship Envision Work Stations
Adjustable height worksurface for your flexibility and productivity
  • Furnished with shelving, electrical fixtures, and work surfaces
  • Available in epoxy or phenolic work surfaces
  • Work surface is adjustable from 30 to 36 in
  • Designed for back to back use for flexible space
Save space in your lab with these top-quality workstations. Choose from a epoxy or phenolic work surface, 48, 60, or 72 in widths, and three or four duplex outlets. The heavy-duty white power coated steel construction is 84 in (213.4 cm) tall and 30 in (76.2 cm) deep. It is supplied with a GFCI protected power, data ports, and three tiers of 12” deep adjustable steel shelving with front and rear retaining rods. Both electrical and plumbing can be ran to either side of the work station from the overhead service panel. The power cord is designed to reach 48 in above the work station, and has a 125 V, 20 amp circuit that includes a twist lock NEMA L5-20P plug. Each post is punched and capped to accept additional plumbing and task lighting.
$5,600.00 - $9,510.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
ICI Quick Ship Envision Workstation, 48" W, Epoxy Work Surface EW-78984-32 Mfr # CPENO-3048E
COMPARE
84 Epoxy work surface 48
$6,365.00USD / Each
ICI Quick Ship Envision Workstation, 60" W, Epoxy Work Surface EW-78984-33 Mfr # CPENO-3060E
COMPARE
84 Epoxy work surface 60
$8,310.00USD / Each
ICI Quick Ship Envision Workstation, 72" W, Epoxy Work Surface EW-78984-34 Mfr # CPENO-3072E
COMPARE
84 Epoxy work surface 72
$9,510.00USD / Each
ICI Quick Ship Envision Workstation, 48" W, Phenolic Work Surface EW-78984-35 Mfr # CPENO-3048P
COMPARE
84 Phenolic work surface 48
$5,600.00USD / Each
ICI Quick Ship Envision Workstation, 60" W, Phenolic Work Surface EW-78984-36 Mfr # CPENO-3060P
COMPARE
84 Phenolic work surface 60
$6,480.00USD / Each
ICI Quick Ship Envision Workstation, 72" W, Phenolic Work Surface EW-78984-37 Mfr # CPENO-3072P
COMPARE
84 Phenolic work surface 72
$7,370.00USD / Each