ICI Quick Ship Stationary Gray Wire Shelving Systems
Create versatile custom shelving to suit your needs
  • Open-wire design allows air circulation to reduce dust accumulation
  • Easily adjust shelves at 1” intervals
  • Available in chrome-plated steel, 304 stainless steel, and Metroseal™ 3 epoxy-coated steel
  • Free standing and mobile options
  • 2000-lb total capacity
Create your innovative storage space with ICI Quick Ship wire shelves. Open-wire shelves are easily adjustable at 1” intervals along the entire height of the post. Choose from three materials: chrome-plated carbon-steel, type 304 polished stainless steel, or corrosion-resistant Metroseal 3 epoxy-coated steel. The Metroseal 3 epoxy with Microban® provides protection from bacteria, mold, mildew, and fungi buildup that may cause odor, stains, and product degradation—ideal for storage in cold damp environments.
Shelving units include four posts, four shelves, split-sleeves to attach shelves to posts, and four adjustable leveling feet.
Shelves that are 36” to 54” have an 800-lb load capacity per shelf while the 60” to 72” long shelves have a 600-lb load capacity per shelf. Shelving unit has a 2000-lb weight capacity with a uniformly distributed load.
Order accessories to customize your shelving system. Use the wire shelf dividers to keep shelf contents organized. The wire shelf ledges prevent items from falling off of shelves. Wire slide 78982-52 easily attaches to the shelf frame and holds the tote boxes. The clear plastic labels holders accept most 1-1/4” labels to quickly identify shelf contents.
$401.00 - $4,750.00USD / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Depth (in) Material Width (in)
Availability Pricing
ICI Quick Ship Stationary Wire Shelving, Chrome; 18" D, 36" W, 74" H EW-78982-04 Mfr # CPJ1836-74C
COMPARE
18 Chrome 36
$401.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Chrome; 18" D, 48" W, 74" H EW-78982-05 Mfr # CPJ1848-74C
COMPARE
18 Chrome 48
$514.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Chrome; 18" D, 60" W, 74" H EW-78982-06 Mfr # CPJ1860-74C
COMPARE
18 Chrome 60
$517.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Chrome; 18" D, 72" W, 74" H EW-78982-07 Mfr # CPJ1872-74C
COMPARE
18 Chrome 72
$580.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Chrome; 24" D, 36" W, 74" H EW-78982-08 Mfr # CPJ2436-74C
COMPARE
24 Chrome 36
$462.50USD / Each
ICI Quick Ship Stationary Wire Shelving, Chrome; 24" D, 48" W, 74" H EW-78982-09 Mfr # CPJ2448-74C
COMPARE
24 Chrome 48
$605.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Chrome; 24" D, 60" W, 74" H EW-78982-10 Mfr # CPJ2460-74C
COMPARE
24 Chrome 60
$689.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Chrome; 24" D, 72" W, 74" H EW-78982-11 Mfr # CPJ2472-74C
COMPARE
24 Chrome 72
$789.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Stainless Steel; 18" D, 36" W, 74" H EW-78982-12 Mfr # CPSS1836
COMPARE
18 Stainless steel 36
$2,200.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Stainless Steel; 18" D, 48" W, 74" H EW-78982-13 Mfr # CPSS1848
COMPARE
18 Stainless steel 48
$2,505.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Stainless Steel; 18" D, 60" W, 74" H EW-78982-14 Mfr # CPSS1860
COMPARE
18 Stainless steel 60
$3,385.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Stainless Steel; 18" D, 72" W, 74" H EW-78982-15 Mfr # CPSS1872
COMPARE
18 Stainless steel 72
$3,885.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Stainless Steel; 24" D, 36" W, 74" H EW-78982-16 Mfr # CPSS2436
COMPARE
24 Stainless steel 36
$2,900.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Stainless Steel; 24" D, 48" W, 74" H EW-78982-17 Mfr # CPSS2448
COMPARE
24 Stainless steel 48
$3,365.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Stainless Steel; 24" D, 60" W, 74" H EW-78982-18 Mfr # CPSS2460
COMPARE
24 Stainless steel 60
$3,950.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Stainless Steel; 24" D, 72" W, 74" H EW-78982-19 Mfr # CPSS2472
COMPARE
24 Stainless steel 72
$4,750.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Epoxy Coated; 18" D, 36" W, 74" H EW-78982-62 Mfr # CPJ1836-74K
COMPARE
18 Epoxy coated 36
$455.50USD / Each
ICI Quick Ship Stationary Wire Shelving, Epoxy Coated; 18" D, 48" W, 74" H EW-78982-63 Mfr # CPJ1848-74K
COMPARE
18 Epoxy coated 48
$514.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Epoxy Coated; 18" D, 60" W, 74" H EW-78982-64 Mfr # CPJ1860-74K
COMPARE
18 Epoxy coated 60
$582.00USD / Each
ICI Quick Ship Stationary Wire Shelving, Epoxy Coated; 18" D, 72" W, 74" H EW-78982-65 Mfr # CPJ1872-74K
COMPARE
18 Epoxy coated 72
$658.00USD / Each
Showing 1 - 20 of 24