ICI Quick Ship Task Lighting
LED light provides up to 50,000 hours of life
  • Ten-hour automatic shut-off save energy
  • Easily attaches to metal cabinets or phenolic shelves
Task lighting units offer state-of the-art, energy-saving components which provide 50,000 hours of operation. Features include a touch-and-hold continuous dimmer and 10-hour automatic shut-off. Constructed of anodized aluminum, these lights can be added to your existing metal cabinets or phenolic shelves. A painted metal strip is included for use with phenolic shelves. It attaches to the underside of the shelf to help receive the magnet mounts. Add on units are available for additional lighting.
$303.50 - $725.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bulb Type Mounting Type Width (in)
Availability Pricing
ICI Quick Ship Task Lighting, Starter Unit, for use with 36" Wide Metal Cabinet EW-78986-07 Mfr # CPLED-36S
COMPARE
LED Magnetic 23.3
$517.00USD / Each
ICI Quick Ship Task Lighting, Starter Unit, for use with 48" Wide Metal Cabinet EW-78986-08 Mfr # CPLED-48S
COMPARE
LED Magnetic 30.1
$631.00USD / Each
ICI Quick Ship Task Lighting, Starter Unit, for use with 24" Wide Phenolic Shelf EW-78986-09 Mfr # CPLED-24S
COMPARE
LED Magnetic 16.4
$652.00USD / Each
ICI Quick Ship Task Lighting, Starter Unit, for use with 30" or 36" Wide Phenolic Shelf EW-78986-10 Mfr # CPLED-3036S
COMPARE
LED Magnetic 23.3
$603.00USD / Each
ICI Quick Ship Task Lighting, Starter Unit, for use with 42" or 48" Wide Phenolic Shelf EW-78986-11 Mfr # CPLED-4248S
COMPARE
LED Magnetic 30.1
$531.00USD / Each
ICI Quick Ship Task Lighting, Starter Unit, for use with 60" Wide Phenolic Shelf EW-78986-12 Mfr # CPLED-60S
COMPARE
LED Magnetic 43.7
$725.00USD / Each
ICI Quick Ship Task Lighting, Add On Unit, for use with 36" Wide Metal Cabinet EW-78986-13 Mfr # CPLED-36AO
COMPARE
LED Magnetic 23.3
$373.50USD / Each
ICI Quick Ship Task Lighting, Add On Unit, for use with 48" Wide Metal Cabinet EW-78986-14 Mfr # CPLED-48AO
COMPARE
LED Magnetic 30.1
$461.50USD / Each
ICI Quick Ship Task Lighting, Add On Unit, for use with 24" Wide Phenolic Shelf EW-78986-15 Mfr # CPLED-24AO
COMPARE
LED Magnetic 16.4
$303.50USD / Each
ICI Quick Ship Task Lighting, Add On Unit, for use with 30" or 36" Wide Phenolic Shelf EW-78986-16 Mfr # CPLED-3036AO
COMPARE
LED Magnetic 23.3
$303.50USD / Each
ICI Quick Ship Task Lighting, Add On Unit, for use with 42" or 48" Wide Phenolic Shelf EW-78986-17 Mfr # CPLED-4248AO
COMPARE
LED Magnetic 30.1
$425.00USD / Each
ICI Quick Ship Task Lighting, Add On Unit, for use with 60" Wide Phenolic Shelf EW-78986-18 Mfr # CPLED-60AO
COMPARE
LED Magnetic 43.7
$573.00USD / Each
Showing 1- 12 of 12