Mettler Toledo EasyPlus Visible and UV/Visible Spectrophotometers
3-in-1 functionality saves bench space and reduces your investment in equipment and service
  • Conventional spectrophotometry, color measurements, and water analyses can be performed using one instrument
  • Exchangeable XPathHolder™ cuvette carousels allow you to shift quickly between seven different path lengths
  • Avoid errors — PathDetect™ securely checks the path length so your results are accurate
  • Make setup easy with the user-friendly 4.3” touchscreen and interface with workflow guidance and integrated tutorials
  • SmartLid™ automatically starts the measurement, increases productivity, and avoids time-consuming task repetition
The Mettler Toledo EasyPlus spectrophotometers combine conventional spectrophotometry, color measurements, and water analyses in one instrument. Built-in methods eliminate calculation and transcription errors and are easily accessed with built-in features such as a favorite list and LongClick™. Photometric data is automatically calculated using pre-programmed methods to obtain results quickly. Methods that are compliant to standards such as ASTM, DIN, ISO, and EPA eliminate the need for post-measurement calculation and data transcription to achieve accurate results efficiently.
Color measurements are obtained from absorbance or transmittance measurements converted to a color value according to the selected color scale. 30 color scales are available including CIELAB, APHA, Gardner, and more. Water analysis is performed with the SQKitReader™ accessory, sold separately as item 83700-80. SQKitReader™ barcode reader scans Spectroquant™ water test kit barcodes to automatically start methods inputted on the user-friendly touchscreen.
$4,630.00 - $6,915.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bandwidth Min Wavelength (nm) Source Lamp
Availability Pricing
Mettler Toledo 30705169 Easy UV UV/Visible Spectrophotometer; 100-240 VAC, 50/60 Hz EW-83700-04 Mfr # 30705169
COMPARE
3 nm 190 Xenon flash lamp
$6,915.00USD / Each
Mettler Toledo 30705170 Easy VIS Visible Spectrophotometer; 100-240 VAC, 50/60 Hz EW-83700-05 Mfr # 30705170
COMPARE
4 nm 330 Tungsten lamp
$4,630.00USD / Each