NSI Lab Solutions Environmental Microbiology Drinking Water Proficiency Test Cultures
Ensure state and accreditation regulation compliance with proficiency testing
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.
These environmental microbiology drinking water proficiency tests are designed for quantitative and/or qualitative identification of target microorganisms. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation.
If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
$154.00 - $302.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Organism Packaging
Availability Pricing
COMPARE
Quantitative Coliform Drinking Water Proficiency Test Coliform 2/PK
$160.00USD / Each
COMPARE
Qualitative Enterococci Drinking Water Proficiency Test; 10/PK Enterococci 10/PK
$302.00USD / Pkg of 10
COMPARE
Quantitative/Qualitative Coliform/E. coli Drinking Water Proficiency Test; 10/PK Coliform, E. coli 10/PK
$302.00USD / Pkg of 10
COMPARE
Quantitative Enterococci Drinking Water Proficiency Test Enterococci 2/PK
$164.00USD / Pkg of 2
COMPARE
Quantitative Standard Plate Count Drinking Water Proficiency Test Enterococci 2/PK
$154.00USD / Each