NSI Lab Solutions Environmental Microbiology Wastewater Proficiency Test Cultures
Ensure state and accreditation regulation compliance with proficiency testing
  • Monthly testing without order deadlines
  • Convenient online reporting with PT Datalink
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.
These environmental microbiology wastewater proficiency tests are designed for quantitative analysis of target microorganisms. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation.
If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
$164.00 - $333.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Organism
Availability Pricing
COMPARE
Quantitative Coliforms/E.coli Wastewater Proficiency Test Coliforms, E.coli
$164.00USD / Each
COMPARE
Quantitative Fecal Coliform in Sludge Wastewater Proficiency Test Fecal Coliform
$185.00USD / Each
COMPARE
Quantitative Enterococcus/Fecal Strep Wastewater Proficiency Test Enterococcus, Fecal Strep
$164.00USD / Each
COMPARE
Quantitative Standard Plate Count Wastewater Proficiency Test Enterococci
$164.00USD / Each
COMPARE
Quantitative Legionella Wastewater Proficiency Test Legionella
$333.00USD / Each