NSI Lab Solutions Environmental Microbiology Wastewater PT Express™ Cultures
On-demand proficiency test (PT) for when you need results fast
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.
These environmental microbiology wastewater proficiency tests are designed for quantitative analysis of target microorganisms. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation.
Environmental Microbiology Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
$206.00 - $390.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Quantitative Fecal Coliform in Sludge Wastewater PT Express™
$206.00USD / Each
COMPARE
Quantitative Standard Plate Count Wastewater PT Express™
$243.00USD / Each
COMPARE
Quantitative Legionella Wastewater PT Express™
$390.00USD / Each
COMPARE
Quantitative Enterococcus/Fecal Strep Wastewater PT Express™
$243.00USD / Each
COMPARE
Quantitative Coliforms/E.coli/Fecal Coliform Wastewater PT Express™
$243.00USD / Each