NSI Lab Solutions Environmental Wastewater Certified Reference Materials
Validate analytical methods and evaluate the quality of your results with certified reference materials
  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.
$56.00 - $341.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
COMPARE
Orthophosphate-P QC CRM 1/EA 21
$62.00USD / Each
COMPARE
Hardness QC CRM 1/EA 250
$83.00USD / Each
COMPARE
Hardness QC CRM 4/PK 250
$293.00USD / Pkg of 4
COMPARE
Turbidity QC CRM 1/EA 500
$96.00USD / Each
COMPARE
Turbidity QC CRM 4/PK 500
$300.00USD / Pkg of 4
COMPARE
Simple Nutrients QC CRM 1/EA 500
$56.00USD / Each
COMPARE
Complex Nutrients QC CRM 1/EA 500
$57.00USD / Each
COMPARE
Minerals QC CRM 1/EA 500
$95.00USD / Each
COMPARE
Minerals QC CRM 4/PK 500
$341.00USD / Pkg of 4
Showing 1- 9 of 9