NSI Lab Solutions Food Chemistry Allergen Reference Standards
Validate and evaluate samples for common food allergens with quality reference standards
  • Improve safety with blank, high, and low reference materials
  • Validate the accuracy of your allergen testing methods
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 and ISO 9001 requirements
Food allergen reference material standards are used primarily for quality control and validation purposes in food testing applications including calibration, validation, and quality control testing. These reference materials are crucial in demonstrating testing procedures are sensitive and specific enough to detect allergens at levels relevant to consumer safety. By testing reference materials with known allergen concentrations, labs can verify the accuracy, precision, and reliability of their methods. Allergens are available in blank, high, and low concentrations.
$348.00 - $452.00USD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Organism Product Type Weight g (g)
Availability Pricing
COMPARE
Gluten Blank 50
$348.00USD / Each
COMPARE
Gluten Low Level 50
$452.00USD / Each
COMPARE
Gluten High Level 50
$452.00USD / Each
COMPARE
Soy Blank 50
$351.00USD / Each
COMPARE
Peanut Blank 50
$351.00USD / Each
COMPARE
Egg Blank 50
$350.00USD / Each
COMPARE
Total Milk Blank 50
$350.00USD / Each
COMPARE
Egg Low Level 50
$405.00USD / Each
COMPARE
Crustacean High Level 50
$407.00USD / Each
COMPARE
Crustacean Low Level 50
$407.00USD / Each
COMPARE
Soy High Level 50
$405.00USD / Each
COMPARE
Soy Low Level 50
$405.00USD / Each
COMPARE
Peanut High Level 50
$410.00USD / Each
COMPARE
Peanut Low Level 50
$407.00USD / Each
COMPARE
Total Milk High Level 50
$405.00USD / Each
COMPARE
Total Milk Low Level 50
$405.00USD / Each
Showing 1- 16 of 16