NSI Lab Solutions Microbiological Environmental Mixed Cultures
Multiple organism CRM covers six methods in a single pellet!
  • Hydration fluid dissolves culture pellet in less than a minute
  • Compatible with MTF, IDEXX, and plate count methods—CoA includes expected value for multiple methods
  • Less than 10% RSD lot precision
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • No royalty fee
These microbiological reference materials are designed for quality control of water samples. Each dehydrated pellet contains multiple microorganisms that can be tested at once. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Values listed below are approximate activity levels; see the Certificate of Analysis (CoA) for actual values. Store in the freezer.
Microbiology Tests at Approximate Activity Levels:
Total Coliform: 2400 CFU/100 mL
E. coli: 1000 CFU/100 mL
Fecal Coliform: 500 CFU/100 mL
P. aeruginosa: 1000 CFU/100 mL
Enterococci: 1000 CFU/100 mL
HPC: 50 CFU/mL
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
$207.00 - $321.00USD / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Packaging
Availability Pricing
COMPARE
10/PK
$207.00USD / Pkg of 10
COMPARE
20/PK
$321.00USD / Pkg of 20