NSI Lab Solutions Microbiological Food and AgriCultures Mixed Cultures
Multi-organism Certified Reference Material (CRM) for food safety testing
  • Compatible with 3M™ Petrifilm™ plate
  • Hydration fluid dissolves culture pellet in less than a minute
  • No more than 2 passages from primary culture
  • ISO 17034 accredited
  • No royalty fee
Use these microbiological reference materials for quality control of food and agriculture samples. Each dehydrated pellet packaged in a vacuum-sealed glass vial contains multiple microorganisms that can be tested at once. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. These CRMs are designed for use with Petrifilm™ plate, FDA Bacteriological Analytical Manual (BAM) methods, or other re-hydratable film methods. Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
$234.00 - $433.00USD / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description Packaging
Availability Pricing
COMPARE
Multi-Organism CRM with Yeast/Mold; 10/PK 10/PK
$234.00USD / Pkg of 10
COMPARE
Multi-Organism CRM with Yeast/Mold; 20/PK 20/PK
$345.00USD / Pkg of 20
COMPARE
Multi-Organism CRM with Yeast/Mold/B. cereus; 10/PK 10/PK
$294.00USD / Pkg of 10
COMPARE
Multi-Organism CRM with Yeast/Mold/B. cereus; 20/PK 20/PK
$433.00USD / Pkg of 20
COMPARE
Multi-Organism CRM with Yeast; 20/PK 20/PK
$345.00USD / Pkg of 20
COMPARE
Multi-Organism CRM with Yeast; 10/PK 10/PK
$234.00USD / Pkg of 10