NSI Lab Solutions Microbiological Pharmaceutical Cultures, 10K CFU
A single pellet is formulated to recover approximately 10,000 CFU
  • Reliable and easy to use – 10,000 CFU per pellet
  • Hydration fluid dissolves culture pellet in less than a minute
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
These microbiological reference materials are designed for use as quantitative control for pharmaceutical culture analysis. The single strain ISO 17034 certified reference material is formulated to recover approximately 10,000 CFU/pellet by cultural plating techniques. For your convenience, each pack includes ten culture pellets individually vacuum sealed in 5 mL glass vials. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Rehydration of a single 10,000 CFU/pellet in 100 mL fluid would provide 100 CFU per mL. Following this prescribed rehydration scheme provides up to 100 replicate tests per hydrated pellet. Extra activity listed on Certificate of Analysis (CoA). Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
$375.00USD / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Organism Organism type
Availability Pricing
COMPARE
S. enterica Abony NCTC 6017, WDCM 00029
$375.00USD / Pkg of 10
COMPARE
B. subtilis NCTC 10400, WDCM 00003, ATCC 6633
$375.00USD / Pkg of 10
COMPARE
C. albicans NCPF 3179, WDCM 00054, ATCC 10231
$375.00USD / Pkg of 10
COMPARE
A. brasiliensis NCPF 2275, WDCM 00053, ATCC 16404
$375.00USD / Pkg of 10
COMPARE
E. coli NCTC 12933, WDCM 0012, ATCC 8739
$375.00USD / Pkg of 10
COMPARE
S. aureus NCTC 10788, WDCM 0032, ATCC 6538
$375.00USD / Pkg of 10